Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029723.3 | NP_083999.1 | 4 | 343 | ||
GenBank mRNA | AK009567.1 | - | 4 | 340 | ||
AK133382.1 | - | 459 | ||||
BC052745.1 | - | 478 | ||||
BC063096.1 | - | 5 | 435 | |||
BC116916.1 | - | 4 | 333 | 40126235 | ||
BC116920.1 | - | 4 | 333 | 40126239 |