Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsdma2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029727.2 | NP_084003.2 | 4 | 465 | ||
GenBank mRNA | AK008663.1 | - | 4 | 462 | ||
AK008685.1 | - | 5 | 497 | |||
BC109335.1 | - | 3 | 368 | 40052139 | ||
BC109336.1 | - | 4 | 399 | 40052146 |