Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtif2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282118.1 | NP_001269047.1 | 12, 13 | 2049 | ||
NM_001282119.1 | NP_001269048.1 | 12, 13 | 1971 | |||
NM_001282120.1 | NP_001269049.1 | 12, 13 | 2413 | |||
NM_133767.3 | NP_598528.2 | 12, 13 | 2327 | |||
GenBank mRNA | AK136344.1 | - | 12, 13 | 2011 | ||
AK146961.1 | - | 12, 13 | 2267 | |||
BC016590.1 | - | 12, 13 | 1903 | 3585359 | ||
BC066819.1 | - | 12, 13 | 2307 |