Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mzt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175245.4 | NP_780454.1 | 2, 3 | 284 | ||
GenBank mRNA | AK010788.1 | - | 2, 3 | 260 | ||
AK082817.1 | - | 2, 3 | 230 | |||
AK087564.1 | - | 2, 3 | 228 | |||
AK150268.1 | - | 2, 3 | 260 | |||
AK151232.1 | - | 2, 3 | 260 | |||
AK151358.1 | - | 2, 3 | 260 | |||
AK151819.1 | - | 2, 3 | 260 | |||
AK160911.1 | - | 2, 3 | 269 | |||
AK165605.1 | - | 2, 3 | 247 | |||
AK168114.1 | - | 2, 3 | 225 | |||
AK169036.1 | - | 2, 3 | 234 | |||
AK170746.1 | - | 2, 3 | 234 | |||
AK209030.1 | - | 1, 2 | 110 | |||
BB844588.1 | - | 2, 3 | 285 | |||
BC038168.1 | - | 2, 3 | 222 | |||
BC098481.1 | - | 2, 3 | 247 | |||
BC117005.1 | - | 2, 3 | 259 | 40126324 | ||
BC117007.1 | - | 2, 3 | 259 | 40126326 |