Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2410131K14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081236.1 | NP_001074705.1 | 1 | 201 | ||
GenBank mRNA | AK010791.1 | - | 1 | 178 | ||
AK090380.1 | - | 1 | 160 | |||
BC104372.1 | - | 1 | 135 | 40049898 | ||
BC104373.1 | - | 1 | 135 | 40049899 |