Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bri3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029752.2 | NP_084028.1 | 3 | 573 | ||
GenBank mRNA | AK010810.1 | - | 3 | 516 | ||
AK043288.1 | - | 3 | 515 | |||
AK044565.1 | - | 3 | 461 | |||
AK143538.1 | - | 3 | 532 | |||
BB618776.1 | - | 3 | 563 | |||
BC039941.1 | - | 3 | 531 | 4011957 |