Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nubpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029760.2 | NP_084036.2 | 9 | 836 | ||
GenBank mRNA | AK010827.1 | - | 9 | 818 | ||
AK141257.1 | - | 9 | 800 | |||
AK152377.1 | - | 9 | 837 | |||
BC023294.1 | - | 430 | ||||
BC062175.1 | - | 9 | 817 | 30291107 |