Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nubpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029760.2 | NP_084036.2 | 11 | 981 | ||
GenBank mRNA | AI118524.1 | - | 1 | 246 | ||
AK010827.1 | - | 11 | 963 | |||
AK141257.1 | - | 11 | 945 | |||
AK152377.1 | - | 11 | 982 | |||
BC023294.1 | - | 575 | ||||
BC062175.1 | - | 11 | 962 | 30291107 |