Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164363.1 | NP_001157835.1 | 7, 8 | 1055 | ||
NM_001164365.1 | NP_001157837.1 | 7, 8 | 712 | |||
NM_029810.4 | NP_084086.3 | 7, 8 | 643 | |||
GenBank mRNA | AK008045.1 | - | 7, 8 | 590 | ||
AK030786.1 | - | 8, 9 | 660 | |||
AK036730.1 | - | 7, 8 | 601 | |||
AK038458.1 | - | 4, 5 | 348 | |||
AK132384.1 | - | 7, 8 | 601 | |||
AK147901.1 | - | 7, 8 | 601 | |||
AK162385.1 | - | 7, 8 | 1020 | |||
AK169616.1 | - | 7, 8 | 601 | |||
BC064760.1 | - | 6, 7 | 577 | 30436992 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC006028.1 | - | 946 | Chr. - - [-] on Build |