Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ticrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029835.1 | NP_084111.1 | 19 | 3327 | ||
GenBank mRNA | AK051639.1 | - | 19 | 3290 | ||
AK054492.1 | - | 19 | 3294 | |||
AK137597.1 | - | 19 | 3332 | |||
BC158103.1 | - | 19 | 3291 | 9088180 |