Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem181b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC147312.1 | - | 226 | 9056375 | ||
BC147313.1 | - | 226 | 9056366 |
Gene Symbol: Tmem181a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033178.3 | NP_001028350.1 | 3 | 246 | ||
GenBank mRNA | AK122507.1 | - | 3 | 250 | ||
AK154554.1 | - | 3 | 242 | |||
CD539539.1 | - | 3 | 173 | |||
CJ168748.1 | - | 3 | 246 |