Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164724.1 | NP_001158196.1 | 3 | 249 | ||
NM_133775.2 | NP_598536.2 | 3 | 228 | |||
GenBank mRNA | AK020353.1 | - | 3 | 229 | ||
AK075849.1 | - | 3 | 250 | |||
AK163464.1 | - | 3 | 229 | |||
AY905582.1 | - | 2 | 169 | |||
BC003847.1 | - | 3 | 205 | 3593927 |