Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164724.1 | NP_001158196.1 | 8 | 725 | ||
NM_133775.2 | NP_598536.2 | 8 | 704 | |||
GenBank mRNA | AK020353.1 | - | 8 | 708 | ||
AK075849.1 | - | 8 | 726 | |||
AK163464.1 | - | 8 | 705 | |||
AY905582.1 | - | 7 | 645 | |||
BC003847.1 | - | 8 | 681 | 3593927 | ||
HQ176370.1 | - | 5 | 345 |