Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164724.1 | NP_001158196.1 | 8 | 841 | ||
NM_133775.2 | NP_598536.2 | 8 | 820 | |||
GenBank mRNA | AK020353.1 | - | 8 | 824 | ||
AK075849.1 | - | 8 | 842 | |||
AK163464.1 | - | 8 | 821 | |||
BC003847.1 | - | 8 | 797 | 3593927 | ||
HQ176370.1 | - | 5 | 461 |