Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045513.3 | NP_001038978.3 | 4 | 862 | ||
GenBank mRNA | CB246004.1 | - | 1 | 393 |