Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029896.1 | NP_084172.1 | 8, 9 | 1209 | ||
GenBank mRNA | AK020492.1 | - | 2, 3 | 182 | ||
AK035109.1 | - | 8, 9 | 1211 | |||
AK037620.1 | - | 8, 9 | 1216 | |||
AK149692.1 | - | 8, 9 | 1210 | |||
AK159240.1 | - | 8, 9 | 1209 | |||
BC019115.1 | - | 7, 8 | 735 | |||
BC031502.1 | - | 8, 9 | 899 | |||
BC094419.1 | - | 8, 9 | 1171 | |||
BC157943.1 | - | 1132 | 9007278 | |||
BC157945.1 | - | 1133 | 9007280 | |||
BC158054.1 | - | 1133 | 9007356 | |||
EF011615.1 | - | 8, 9 | 898 |