Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2afv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029938.1 | NP_084214.1 | 4 | 335 | ||
GenBank mRNA | AK021267.1 | - | 4 | 258 | ||
AK155105.1 | - | 4 | 338 | |||
AK168110.1 | - | 4 | 307 | |||
AK178770.1 | - | 2 | 138 | |||
AK187484.1 | - | 3 | 182 | |||
BC028539.1 | - | 3 | 130 | |||
BC147293.1 | - | 3 | 294 | 9056353 | ||
BC147294.1 | - | 4 | 320 | 9056344 | ||
BC147741.1 | - | 4 | 320 | 9056690 | ||
BC147751.1 | - | 4 | 320 | 9056700 | ||
CJ069840.1 | - | 4 | 336 |