Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029965.2 | NP_084241.1 | 2, 3 | 351 | ||
GenBank mRNA | AK020104.1 | - | 2, 3 | 256 | ||
AK036051.1 | - | 2, 3 | 210 | |||
AK044579.1 | - | 357 | ||||
AK080591.1 | - | 2, 3 | 151 | |||
AK152760.1 | - | 2, 3 | 341 | |||
AK153658.1 | - | 2, 3 | 352 | |||
AK162554.1 | - | 2, 3 | 365 | |||
BC031383.1 | - | 2, 3 | 165 | |||
BC114210.1 | - | 2, 3 | 351 | 6309632 | ||
BC138930.1 | - | 2, 3 | 179 | 8861952 | ||
BC145718.1 | - | 2, 3 | 184 | 40131021 |