Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029965.2 | NP_084241.1 | 3, 4 | 487 | ||
GenBank mRNA | AK020104.1 | - | 3, 4 | 392 | ||
AK036051.1 | - | 3, 4 | 346 | |||
AK044579.1 | - | 493 | ||||
AK080591.1 | - | 3, 4 | 287 | |||
AK152760.1 | - | 3, 4 | 477 | |||
AK153658.1 | - | 3, 4 | 488 | |||
AK162554.1 | - | 3, 4 | 502 | |||
BC031383.1 | - | 3, 4 | 301 | |||
BC114210.1 | - | 3, 4 | 487 | 6309632 | ||
BC138930.1 | - | 3, 4 | 315 | 8861952 | ||
BC145718.1 | - | 3, 4 | 320 | 40131021 |