Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029983.5 | NP_084259.1 | 6 | 674 | ||
GenBank mRNA | AF287467.1 | - | 6 | 725 | ||
AF434990.1 | - | 6 | 717 | |||
AK020837.1 | - | 4 | 331 | |||
AK030877.1 | - | 6 | 675 | |||
AK088672.1 | - | 6 | 801 | |||
AK138709.1 | - | 6 | 675 | |||
BC052655.1 | - | 6 | 773 | 30040401 |