Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tchp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029992.2 | NP_084268.2 | 2, 3 | 370 | ||
GenBank mRNA | AK020912.1 | - | 2, 3 | 333 | ||
AK083027.1 | - | 2, 3 | 286 | |||
AK159947.1 | - | 2, 3 | 331 | |||
BC043048.1 | - | 2, 3 | 275 | 5704036 | ||
BU512279.1 | - | 2, 3 | 370 |