Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100451.2 | NP_001093921.1 | 1584 | |||
GenBank mRNA | AK153696.1 | - | 2174 | |||
AK173208.1 | - | 2355 | ||||
BC070432.1 | - | 1584 | ||||
BC100371.1 | - | 1 | 738 |