Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ypel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005341.3 | NP_001005341.1 | 3, 4 | 521 | ||
GenBank mRNA | AB098741.2 | - | 2, 3 | 237 | ||
AK021387.1 | - | 3, 4 | 467 | |||
BC121190.1 | - | 3, 4 | 393 | 6467362 |