Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ypel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005341.3 | NP_001005341.1 | 4 | 570 | ||
GenBank mRNA | AB098741.2 | - | 3 | 286 | ||
BC121190.1 | - | 4 | 442 | 6467362 |