Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grifin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030022.1 | NP_084298.1 | 4 | 365 | ||
GenBank mRNA | AK021365.1 | - | 4 | 366 | ||
BC107323.2 | - | 4 | 365 | 40053455 | ||
BC107324.1 | - | 4 | 365 | 40053456 | ||
BC107343.2 | - | 2 | 179 |