Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cst11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030059.2 | NP_084335.1 | 1 | 300 | ||
GenBank mRNA | AF454373.1 | - | 1 | 302 | ||
AK020300.1 | - | 1 | 302 | |||
BC100526.1 | - | 1 | 225 | 30920256 |