Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030068.1 | NP_084344.1 | 14 | 2022 | ||
GenBank mRNA | AK019535.1 | - | 14 | 2054 | ||
AK029502.1 | - | 14 | 2058 | |||
BC141047.1 | - | 14 | 2022 | 9055659 | ||
BC145231.1 | - | 14 | 2022 | 9053783 |