Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc88b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081291.1 | NP_001074760.1 | 14 | 2284 | ||
GenBank mRNA | BC076600.1 | - | 5 | 1168 | ||
BC151001.1 | - | 14 | 2257 | 9087914 | ||
BC151009.1 | - | 14 | 2257 | 9087922 | ||
BC157946.1 | - | 14 | 2257 | 9088122 |