Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083891.1 | NP_001077360.1 | 3 | 320 | ||
NM_030087.2 | NP_084363.2 | 4 | 1412 | |||
GenBank mRNA | AK077433.1 | - | 3 | 261 | ||
AK154191.1 | - | 4 | 1345 | |||
AK154477.1 | - | 4 | 1391 | |||
AK201972.1 | - | 1 | 64 | |||
BC013640.1 | - | 4 | 1372 | 4188234 | ||
BF318157.1 | - | 1 | 162 | |||
BU555486.1 | - | 3 | 323 | |||
BU755401.1 | - | 1 | 179 |