Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgpep1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030101.1 | NP_084377.1 | 3 | 301 | ||
GenBank mRNA | AK021221.1 | - | 3 | 302 | ||
BY728828.1 | - | 3 | 300 |