Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micu3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030110.1 | NP_084386.1 | 11 | 1165 | ||
GenBank mRNA | BC125419.1 | - | 10 | 759 | ||
BC125445.1 | - | 10 | 759 | |||
BC145173.1 | - | 9 | 720 |