Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042615.2 | NP_001036080.1 | 3 | 979 | ||
NM_030127.3 | NP_084403.2 | 3 | 979 | |||
GenBank mRNA | AK008764.1 | - | 2 | 101 | ||
AK035194.1 | - | 3 | 982 | |||
AY037300.1 | - | 3 | 764 | |||
AY156509.1 | - | 3 | 671 | |||
AY280664.1 | - | 3 | 671 | |||
BC022646.1 | - | 2 | 90 | |||
BC138587.1 | - | 3 | 703 | 8861607 | ||
BC138588.1 | - | 3 | 703 | 8861608 |