Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023898.4 | NP_076387.2 | 2 | 182 | ||
GenBank mRNA | AF189146.1 | - | 1 | 90 | ||
AF190929.1 | - | 1 | 90 | |||
AK020922.1 | - | 2 | 183 | |||
BC057290.1 | - | 154 | ||||
BC058192.1 | - | 2 | 172 | |||
BC115671.1 | - | 2 | 160 | 40104107 | ||
BC115672.1 | - | 2 | 160 | 40104108 |