Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bola3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175277.4 | NP_780486.1 | 3 | 237 | ||
GenBank mRNA | AK019037.1 | - | 2 | 72 | ||
AK028102.1 | - | 3 | 238 | |||
AK131950.1 | - | 246 | ||||
AK160488.1 | - | 3 | 241 | |||
AK218122.1 | - | 1 | 54 | |||
BC049748.1 | - | 3 | 257 | |||
BC116947.1 | - | 4 | 275 | 40126266 | ||
BC116949.1 | - | 4 | 275 | 40126268 |