Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bola3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175277.4 | NP_780486.1 | 2 | 100 | ||
GenBank mRNA | AK028102.1 | - | 2 | 101 | ||
AK131950.1 | - | 109 | ||||
AK160488.1 | - | 2 | 104 | |||
AK213244.1 | - | 1 | 35 | |||
BC049748.1 | - | 2 | 120 | |||
BC116947.1 | - | 2 | 138 | 40126266 | ||
BC116949.1 | - | 2 | 138 | 40126268 |