Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023900.2 | NP_076389.2 | 6 | 919 | ||
GenBank mRNA | AK020621.1 | - | 6 | 588 | ||
AK051734.1 | - | 6 | 575 | |||
AK088567.1 | - | 6 | 882 | |||
AK150144.1 | - | 6 | 534 | |||
AK208036.1 | - | 1 | 14 | |||
BC005565.1 | - | 6 | 517 | 3966305 | ||
BE951415.1 | - | 1 | 354 |