Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc22d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023910.6 | NP_076399.4 | 2 | 1404 | ||
NM_029805.1 | NP_084081.1 | 2 | 1404 | |||
GenBank mRNA | AF201286.1 | - | 2 | 1117 | ||
AK018735.2 | - | 2 | 1377 | |||
AK040534.1 | - | 2 | 1341 | |||
AK079510.1 | - | 2 | 1130 | |||
AK080828.1 | - | 2 | 1419 | |||
AK143608.1 | - | 2 | 1181 | |||
AK144806.1 | - | 1 | 841 | |||
AK150043.1 | - | 2 | 1373 | |||
AK154153.1 | - | 2 | 1354 | |||
AK154490.1 | - | 2 | 1004 | |||
AK155712.1 | - | 2 | 1406 | |||
AK170487.1 | - | 2 | 1393 | |||
BC018544.1 | - | 1 | 876 | 3666221 | ||
BC023761.1 | - | 2 | 1308 | 5343927 | ||
BC145721.1 | - | 2 | 1367 | 40131024 | ||
BC145723.1 | - | 2 | 1367 | 40131026 |