Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc22d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023910.6 | NP_076399.4 | 3 | 1582 | ||
NM_029805.1 | NP_084081.1 | 3 | 1582 | |||
GenBank mRNA | AF201286.1 | - | 3 | 1295 | ||
AK018735.2 | - | 3 | 1555 | |||
AK040534.1 | - | 3 | 1519 | |||
AK079510.1 | - | 3 | 1308 | |||
AK080828.1 | - | 3 | 1597 | |||
AK143608.1 | - | 3 | 1359 | |||
AK144806.1 | - | 2 | 1019 | |||
AK150043.1 | - | 3 | 1551 | |||
AK154153.1 | - | 3 | 1532 | |||
AK154490.1 | - | 3 | 1182 | |||
AK155712.1 | - | 3 | 1584 | |||
AK170487.1 | - | 3 | 1571 | |||
BC018544.1 | - | 2 | 1054 | 3666221 | ||
BC023761.1 | - | 3 | 1486 | 5343927 | ||
BC145721.1 | - | 3 | 1545 | 40131024 | ||
BC145723.1 | - | 3 | 1545 | 40131026 |