Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wrnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030215.3 | NP_084491.3 | 4 | 1606 | ||
GenBank mRNA | AB056151.1 | - | 4 | 1455 | ||
AF208046.1 | - | 4 | 851 | |||
AK041886.1 | - | 4 | 1562 | |||
AK050368.1 | - | 4 | 1608 | |||
AK082078.1 | - | 4 | 1521 | |||
AK167570.1 | - | 4 | 1607 | |||
AK170542.1 | - | 4 | 1599 | |||
AK170846.1 | - | 4 | 1587 | |||
AK184555.1 | - | 2 | 261 | |||
BC010482.1 | - | 4 | 1598 | 3492125 | ||
BC058744.1 | - | 4 | 996 |