Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chsy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081328.1 | NP_001074797.1 | 3 | 1552 | ||
GenBank mRNA | AK019528.1 | - | 3 | 539 | ||
AK220519.1 | - | 1439 | ||||
BC151164.1 | - | 3 | 1242 | 9088053 | ||
BC151165.1 | - | 3 | 1242 | 9088054 | ||
BC157985.1 | - | 3 | 1242 | 9088141 |