Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Avl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030235.1 | NP_084511.1 | 10 | 1139 | ||
GenBank mRNA | AK020003.1 | - | 10 | 1116 | ||
AK083605.1 | - | 10 | 1095 | |||
AK122228.1 | - | 10 | 1117 | |||
AK147365.1 | - | 10 | 1141 | |||
BC051177.1 | - | 10 | 1155 | |||
BC117878.1 | - | 10 | 1101 | 40095961 | ||
BC117879.1 | - | 10 | 1101 |