Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019730.2 | NP_062704.2 | 3 | 248 | ||
GenBank mRNA | AF153449.1 | - | 3 | 260 | ||
AF288689.1 | - | 3 | 242 | |||
AK002525.1 | - | 3 | 228 | |||
AK007399.1 | - | 3 | 248 | |||
BC028503.1 | - | 3 | 239 | 3326648 |