Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023680.3 | NP_076169.2 | 1 | 24 | ||
GenBank mRNA | AK012838.1 | - | 1 | 25 | ||
AY046551.1 | - | 1 | 23 | |||
AY165626.1 | - | 1 | 47 | |||
AY165627.1 | - | 1 | 47 | |||
BC076592.1 | - | 1 | 38 |