Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024468.2 | NP_077788.2 | 2 | 225 | ||
NM_178281.1 | NP_840065.1 | 2 | 210 | |||
GenBank mRNA | AB046382.1 | - | 2 | 225 | ||
AK053507.1 | - | 2 | 162 | |||
AK053591.1 | - | 1 | 3298 | |||
AK145861.1 | - | 2 | 161 | |||
AK220298.1 | - | 2 | 146 | |||
BC031540.1 | - | 1 | 267 | 3666785 | ||
BY312131.1 | - | 2 | 212 |