Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170552.1 | NP_001164023.1 | 16 | 2407 | ||
NM_030675.3 | NP_109600.2 | 17 | 2478 | |||
GenBank mRNA | AF306509.1 | - | 17 | 2173 | ||
AF310134.1 | - | 17 | 2454 | |||
AK041574.1 | - | 5 | 493 | |||
AK085107.1 | - | 16 | 3899 | |||
AK144667.1 | - | 17 | 2471 | |||
AY328895.1 | - | 15 | 2043 | |||
AY464945.1 | - | 16 | 2160 | |||
BC016087.1 | - | 17 | 2426 | |||
BC044820.1 | - | 17 | 2367 | |||
BC054819.1 | - | 18 | 2717 | |||
BC085252.1 | - | 16 | 2167 | |||
BC141042.1 | - | 17 | 2322 | 9055654 |