Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK086088.1 | - | 7 | 1545 | ||
AK122543.1 | - | 5 | 4749 | |||
AK144928.1 | - | 7 | 1544 | |||
AK150129.1 | - | 7 | 1524 | |||
AK156171.1 | - | 7 | 1585 | |||
AK159202.1 | - | 7 | 1576 | |||
AK166491.1 | - | 7 | 1545 | |||
AK167203.1 | - | 7 | 1661 | |||
AK172333.1 | - | 7 | 1585 | |||
AK215492.1 | - | 1 | 137 | |||
BC003251.1 | - | 6 | 1206 | 3500047 | ||
BC013661.1 | - | 3 | 677 | |||
BC094418.1 | - | 7 | 1567 | 30702568 |