Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030251.3 | NP_084527.2 | 10 | 1101 | ||
GenBank mRNA | AB053477.1 | - | 10 | 1032 | ||
AI117801.1 | - | 3 | 272 | |||
AK049100.1 | - | 10 | 997 | |||
AK088437.1 | - | 10 | 1066 | |||
AK089474.1 | - | 7 | 1624 | |||
AK155432.1 | - | 5 | 2523 | |||
AK155505.1 | - | 5 | 2523 | |||
AK155644.1 | - | 5 | 2523 | |||
AK220208.1 | - | 10 | 945 | |||
BC003234.1 | - | 10 | 1045 | 3483893 |