Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apobec3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160415.1 | NP_001153887.1 | 6 | 939 | ||
NM_030255.3 | NP_084531.2 | 5 | 840 | |||
GenBank mRNA | AB461446.1 | - | 6 | 830 | ||
AB461447.1 | - | 5 | 731 | |||
AB461448.1 | - | 5 | 731 | |||
AB461449.1 | - | 5 | 731 | |||
AK044394.1 | - | 6 | 899 | |||
AK049256.1 | - | 5 | 800 | |||
AK049998.1 | - | 794 | ||||
AK153719.1 | - | 5 | 790 | |||
AK154035.1 | - | 944 | ||||
AK167986.1 | - | 5 | 846 | |||
BC003314.1 | - | 6 | 921 | 3155422 | ||
BC007182.1 | - | 2 | 336 | |||
EU707568.1 | - | 6 | 830 | |||
EU707569.1 | - | 5 | 662 | |||
EU707570.1 | - | 5 | 731 | |||
EU707571.1 | - | 5 | 662 | |||
GQ871501.1 | - | 6 | 827 | |||
GQ871502.1 | - | 4 | 671 | |||
GQ871503.1 | - | 5 | 728 |