Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024954.1 | NP_001020125.1 | 3 | 386 | ||
GenBank mRNA | AJ300183.1 | - | 3 | 314 |