Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030724.3 | NP_109649.1 | 7 | 1024 | ||
GenBank mRNA | AF236636.1 | - | 7 | 1029 | ||
AK051894.1 | - | 7 | 794 | |||
AK053745.1 | - | 7 | 984 | |||
AK077517.1 | - | 7 | 963 | |||
AK082837.1 | - | 7 | 987 | |||
AK155939.1 | - | 7 | 988 | |||
AK168318.1 | - | 7 | 1002 | |||
AK191847.1 | - | 1 | 27 | |||
AK193626.1 | - | 1 | 27 | |||
AK193847.1 | - | 1 | 27 | |||
AK194288.1 | - | 1 | 27 | |||
AK197462.1 | - | 1 | 27 | |||
AK199301.1 | - | 1 | 27 | |||
BC023789.1 | - | 7 | 988 | 5352192 |